Would you like to log in again?
Merit™ Sterile Iris Scissors, Disposable, Sklar
Supplier: Sklar
REVIEWS (0)
Total Ratings: 0
Avg. Ratings: out of 5
Sort all by:
Merit™
96-2506M 96-2507M 96-2505M 96-2504M
10022-486CS 922.4 USD InStock
10022-486 10022-580 75995-378 75995-380
Merit™ Sterile Iris Scissors, Disposable, Sklar
Scissors
Merit™ Sterile Iris Scissors were originally designed for ophthalmic procedures, but have become a multipurpose instrument used in various surgical environments.
  • Ready-to-use
  • Multipurpose
  • Ideal for detailed work
These fine, sharp scissors are ideal for detailed dissection of fine tissue and fine suture removal. In addition to ophthalmic procedures, Iris Scissors are now often used multiple procedures. Merit™ Sterile instruments are ready-to-use, sterile, disposable, office-grade instruments sold in bulk quantities.
Order Now
ORDER
WRITE A REVIEW
REVIEWS (0)
Q and A
Product Questions and Answers
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.
Original Product:
Description Catalog Number Availability Unit Your Price Price Per Qty