Would you like to log in again?
Accucore™ Vanquish™ 1.5 µm UHPLC Column, Thermo Scientific
Supplier: Thermo Scientific
Accucore™ Vanquish™
27101-102130 27101-052130 27101-152130
10805-176EA 1938.66 USD InStock
10805-176 10022-582 10805-174
Accucore™ Vanquish™ 1.5 µm UHPLC Column, Thermo Scientific
Chromatography Columns
This sub-2 µm particle, next-generation column has ultra-short diffusion paths that result in extremely efficient separations.
  • More results with better separations
  • Resolution of complex mixtures
  • Ultra-short diffusion paths
Choose Thermo Scientific Accucore Vanquish C18 UHPLC columns, containing uniquely efficient 1.5 µm solid core particles for use with the Thermo Scientific Vanquish UHPLC system. Founded on state-of-the-art Core Enhanced Technology, this sub-2 µm particle next-generation column has ultra-short diffusion paths that result in extremely efficient separations. This high efficiency enables fast separation and the resolution of complex mixtures. In concert with Vanquish systems, these columns deliver more results with better separations, thereby enhancing workflows and productivity.
Order Now
ORDER
SPECIFICATIONS
DOCUMENTATION
Q and A
Product Questions and Answers
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.
Original Product:
Description Catalog Number Availability Unit Your Price Price Per Qty