Would you like to log in again?
Creatinine Enzymatic Reagent Set, Pointe Scientific
Supplier: Pointe Scientific
C7548-1L C7548-480
10022-642EA 528.3 USD InStock
10022-642 10022-644
Creatinine Enzymatic Reagent Set, Pointe Scientific
Assays Enzyme Assays
The Creatinine Enzymatic Reagent Set is for the quantitative enzymatic determination of creatinine in serum and urine.
  • Quantitative enzymatic determination of creatinine in serum and urine
  • Employs a two reagent system
  • Eliminates interference by endogenous creatine and ascorbic acid
The Creatinine Enzymatic Reagent Set is for the quantitative enzymatic determination of creatinine in serum and urine. For In Vitro Diagnostic Use Only. Creatinine is a catabolic product of creatine, which is used in skeletal muscle contraction. The daily production of creatine, and subsequently creatinine, depends on muscle mass, which fluctuates very little. Creatinine is excreted entirely by the kidneys and therefore is directly proportional to renal excretory function. Thus with normal renal excretory function, the serum creatinine level should remain constant and normal. Only renal disorders, such as glomerulonephritis, pyelonephritis, acute tubular necrosis, and urinary obstruction, will cause an abnormal elevation in creatinine. The Pointe Scientific, Inc. method employs a two reagent system which eliminates interference by endogenous creatine and ascorbic acid.
Certifications: CE Certified
Ordering information: Materials Required but not Provided:
Creatinine Standard or Chemistry Calibrator and two levels of control material with known Creatinine levels.
Order Now
ORDER
Q and A
Product Questions and Answers
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.
Original Product:
Description Catalog Number Availability Unit Your Price Price Per Qty