Would you like to log in again?
Triglycerides - Liquid Reagent Set, Pointe Scientific
Supplier: Pointe Scientific
T7532-120 T7532-500 HT932-780 T7532-1L HT932-1000
10023-398EA 1358.4 USD InStock
10023-398 10022-900 10022-902 10023-406 10022-964
Triglycerides - Liquid Reagent Set, Pointe Scientific
Assays Enzyme Assays
Triglycerides - Liquid Reagent Set is used for the in vitro quantitative determination of Triglycerides in serum or plasma.
  • In vitro quantitative determination of Triglycerides in serum or plasma
  • Intensity of the colored complex formed is directly proportional to the triglycerides concentration
  • Diagnostic use only
The Hitachi 780 mL reagent is for use on the Hitachi 917 analyzer only and for in vitro diagnostic use only. There are (12) 65 mL Reagent bottles in this kit.
The Hitachi 1 L reagent is for use on Hitachi 717, 911, 912, and 914 analyzers and for in vitro diagnostic use only. There are (10) 100 mL Reagent bottles in this kit.
The Olympus reagent is for use on the Beckman Coulter AU 400, 480, and 640 analyzers and is for in vitro diagnostic use only. There are (12) 65 mL Reagent bottles in this kit.
Certifications: CE Certified
Ordering information: Materials Required but not Provided:
An NIST-traceable Triglycerides standard or serum calibrator and two levels of Commercially available controls.
Order Now
ORDER
Q and A
Product Questions and Answers
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.
Original Product:
Description Catalog Number Availability Unit Your Price Price Per Qty