Would you like to log in again?
Total Bilirubin (Hitachi and Beckman) Reagent Set, Pointe Scientific
Supplier: Pointe Scientific
HB979-711 OB979-711
10023-470EA 817.07 USD InStock
10023-470 10022-940
Total Bilirubin (Hitachi and Beckman) Reagent Set, Pointe Scientific
Assays Enzyme Assays
The Total Bilirubin (Hitachi & Beckman) Reagent Set is for the quantitative determination of total bilirubin in serum.
  • Quantitative determination of total bilirubin in serum
  • In vitro diagnostic use only
  • The intensity of the color of the azobilirubin produced can be measured photometrically.
The Total Bilirubin (Hitachi & Beckman) Reagent Set is for the quantitative determination of total bilirubin in serum
The Hitachi 711 mL reagent is for use on the Hitachi 917 analyzer only and for in vitro diagnostic use only. There are nine 63 mL R1 Reagent bottles and nine 16 mL R2 Reagent bottles in this kit.
The Hitachi 639 mL reagent is for use on Hitachi 717, 911, 912, and 914 analyzers and for in vitro diagnostic use only. There are six 90 mL R1 Reagent bottles and three 51 mL R2 Reagent bottles in this kit.
The Olympus reagent is for use on the Beckman Coulter AU 400, 480, and 640 analyzers and is for in vitro diagnostic use only. There are nine 63 mL R1 Reagent bottles and nine 16 mL R2 Reagent bottles in this kit.
Certifications: CE Certified
Ordering information: Materials Required but not Provided:
Control sera with known normal and abnormal direct bilirubin values as well as Calibrator.
Order Now
ORDER
DOCUMENTATION
Q and A
Product Questions and Answers
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.
Original Product:
Description Catalog Number Availability Unit Your Price Price Per Qty