Would you like to log in again?
Calcium (Hitachi & Beckman) Reagent Set, Pointe Scientific
Supplier: Pointe Scientific
OC902-576 HC902-576 HC902-1000
10022-992EA 485.26 USD InStock
10022-992 10022-944 10023-476
Calcium (Hitachi & Beckman) Reagent Set, Pointe Scientific
Assays Enzyme Assays
The Calcium Reagent Set is for the quantitative determination of Calcium in serum.
  • Quantitative determination of Calcium in serum
The Calcium Reagent Set is for the quantitative determination of Calcium in serum
The Hitachi 576 mL reagent is for use on the Hitachi 917 analyzer only. There are six 66 mL R1 Reagent bottles and six 30 mL R2 Reagent bottles in this kit.
The Hitachi 1L reagent is for use on Hitachi 717, 911, 912, and 914 analyzers. There are seven 100 mL R1 Reagent bottles and three 100 mL R2 Reagent bottles in this kit.
Certifications: CE Certified
Ordering information: Materials Required but not Provided:
Control sera with known normal and abnormal calcium values as well as Calibrator or Standard.
Order Now
ORDER
Q and A
Product Questions and Answers
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.
Original Product:
Description Catalog Number Availability Unit Your Price Price Per Qty