Would you like to log in again?
Uric Acid Reagent Set, Pointe Scientific
Supplier: Pointe Scientific
U7581-1L U7581-500
10022-970EA 294.51 USD InStock
10022-970 10022-972
Uric Acid Reagent Set, Pointe Scientific
Assays Enzyme Assays
Uric Acid Reagent Set is used for the quantitative determination of Uric Acid in serum.
  • Quantitative determination of Uric Acid in serum
  • For in vitro diagnostic use only
  • Absorbance at 520nm is proportional to the concentration of Uric Acid in the sample
Uric Acid Reagent Set is used for the quantitative determination of Uric Acid in serum. For in vitro diagnostic use only. Uric Acid is oxidized by Uricase to allantoin and hydrogen peroxide. HDCBS + 4-AAP + hydrogen peroxide, in the presence of peroxidase, produces a red chromagen that is measured at 520nm. The absorbance at 520nm is proportional to the concentration of Uric Acid in the sample.
Order Now
ORDER
Q and A
Product Questions and Answers
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.
Original Product:
Description Catalog Number Availability Unit Your Price Price Per Qty