Would you like to log in again?
BUN (Urea Nitrogen) Reagent Set, Pointe Scientific
Supplier: Pointe Scientific
B7552-625 OB952-482 HB952-1020
10022-990EA 574.7 USD InStock
10022-990 10023-472 10022-636
BUN (Urea Nitrogen) Reagent Set, Pointe Scientific
Assays Enzyme Assays
BUN (Urea Nitrogen) Reagent Set is used for the quantitative determination of urea nitrogen in serum.
  • Quantitative determination of urea nitrogen in serum
  • in vitro diagnostic use only
  • Liberated ammonia reacts with α-ketoglutarate in the presence of NADH to yield glutamate
BUN (Urea Nitrogen) Reagent Set is used for the quantitative determination of urea nitrogen in serum
Hitachi BUN 654 ml reagent is for use on the Hitachi 917 analyzer only and for in vitro diagnostic use only. There are six 66 mL R1 Reagent bottles and six 43 mL R2 Reagent bottles in this kit.
Hitachi BUN 1L reagent is for use on Hitachi 717, 911, 912, and 914 analyzers and for in vitro diagnostic use only. There are eight 100 mL R1 Reagent bottles and four 50 mL R2 Reagent bottles in this kit.
Certifications: CE Certified
Ordering information: Materials Required but not Provided:
An NIST-traceable BUN standard (B7550-STD) or serum calibrator (C7506-50) as well as a control sera with known normal and abnormal BUN values (C7590/91/92).
Order Now
ORDER
Q and A
Product Questions and Answers
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.
Original Product:
Description Catalog Number Availability Unit Your Price Price Per Qty