Would you like to log in again?
Creatine Kinase Liquid Reagent Set, Pointe Scientific
Supplier: Pointe Scientific
C7522-625 HC922-250 HC922-300 C7522-150 C7522-450 OC922-300
10022-750EA 973.74 USD InStock
10022-750 10023-480 10022-948 10022-746 10022-748 10022-996
Creatine Kinase Liquid Reagent Set, Pointe Scientific
Assays Enzyme Assays
The Creatine Kinase Liquid Reagent Set is for the quantitative determination of creatine kinase activity in serum.
  • Quantitative determination of creatine kinase activity in serum
  • Provides a definitive diagnosis of acute myocardial infarction
  • For in vitro diagnostic use only
The Creatine Kinase Liquid Reagent Set is for the quantitative determination of creatine kinase activity in serum
The Hitachi 250mL reagent kit is for use on the Hitachi 917 analyzer only and for in vitro diagnostic use only. There are four 60 mL R1 Reagent bottles and four 15 mL R2 Reagent bottles in this kit.
The Hitachi 300 mL reagent kit is for use on Hitachi 717, 911, 912, and 914 analyzers and for in vitro diagnostic use only. There are two 100 mL R1 Reagent bottles and two 25 mL R2 Reagent bottles in this kit.
Certifications: CE Certified
Ordering information: Materials Required but not Provided:
The validity of the reaction should be monitored by use of control sera with known normal and abnormal creatine kinase values (C7590/91/92).
Order Now
ORDER
Q and A
Product Questions and Answers
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.
Original Product:
Description Catalog Number Availability Unit Your Price Price Per Qty