Would you like to log in again?
Super Broth Bottled Media, Teknova
Supplier: Teknova
S0130 S0130
100220-222EA 116.09 USD InStock
100220-222 CA100220-222
Super Broth Bottled Media, Teknova
Media for Microbiology Prepared Media for Microbiology
Used as a cell growth medium to increase plasmid DNA yield.
  • 1 Liter
  • Sterile
Contains 3.2% Tryptone, 2.0% yeast extract, 0.5% NaCl, 0.99% Pottassium Phosphate, Dibasic, pH 7.5. Ambient shipping conditions. Store at room temperature.
Order Now
ORDER
Q and A
Product Questions and Answers
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.
Original Product:
Description Catalog Number Availability Unit Your Price Price Per Qty