Would you like to log in again?
Single Syringe Infusion Pump
Supplier: Stoelting
53100
100229-906EA 2085.77 USD InStock
100229-906
Single Syringe Infusion Pump
Pumps Syringe Pumps
A basic single syringe, infusion pump which combines precision with simplicity.
  • Single syringe 10 µl to 60 ml, any manufacturer
  • Simple menu-driven setup: Select syringe size from displayed table, Set dispense volume, Set dispense flow rate
  • Easy to read bright, backlight display
  • Dispense volume displayed and has automatic shutoff at completion
  • Volume and flow rate units (µl/hr or ml/hr) are selected automatically based on syringe size
  • Last settings stored in permanent memory
  • Button release of drive nut for easy syringe installation
  • Choice of run/stop after a power interruption
The infusion only syringe pumps are ideal for delivering accurate and precise amounts of fluids for a multitude of syringe pump applications. This includes infusing calibrant into a mass spectrometer or reaction chamber, long term drug infusions to animals and general infusion applications.
KD Scientific infusion only syringe pumps are ideal for delivering accurate and precise amounts of fluids for a multitude of syringe pump applications. This includes infusing calibrant into a mass spectrometer or reaction chamber, long term drug infusions to animals and general infusion applications.
KDS syringe pumps are known for their outstanding reliability and performance. The KDS 100 series syringe pumps provide the most cost-effective solution for infusing fluids.
Advance per microstep: 0.529 µ (1/2 step), Max Step Rate (1/2 Step): 400 steps/sec., Min Step Rate: 1 step/30 sec.
Order Now
ORDER
SPECIFICATIONS
Q and A
Product Questions and Answers
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.
Original Product:
Description Catalog Number Availability Unit Your Price Price Per Qty