Would you like to log in again?
Gamma Glutamyl Transferase (Hitachi & Beckman) Reagent Set, Pointe Scientific
Supplier: Pointe Scientific
HG959-315 OG959-295 HG959-295
10023-484EA 509.03 USD InStock
10023-484 10023-390 10022-952
Gamma Glutamyl Transferase (Hitachi & Beckman) Reagent Set, Pointe Scientific
Assays Enzyme Assays
The Gamma Glutamyl Transferase (Hitachi & Beckman) Reagent Set is for the quantitative kinetic determination of gamma glutamyl transferase (GGT) activity in serum.
  • Quantitative kinetic determination of gamma glutamyl transferase (GGT) activity in serum
  • Amount of 5-amino-2-nitrobenzoate formed is proportional to γ-GT activity and may be measured kinetically
The Gamma Glutamyl Transferase (Hitachi & Beckman) Reagent Set is for the quantitative kinetic determination of gamma glutamyl transferase (GGT) activity in serum
The Hitachi 295 mL reagent is for use on the Hitachi 917 analyzer only and for in vitro diagnostic use only. There are five 42 mL R1 Reagent bottles and five 17 mL R2 Reagent bottles in this kit.
The Hitachi 315 mL reagent is for use on Hitachi 717, 911, 912, and 914 analyzers and for in vitro diagnostic use only. There are five 45 mL R1 Reagent bottles and two 45 mL R2 Reagent bottles in this kit.
Certifications: CE Certified, Heath Canada (295 mL Reagent only)
Ordering information: Materials Required but not Provided:
Serums with known normal and abnormal GGT values.
Order Now
ORDER
Q and A
Product Questions and Answers
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.
Original Product:
Description Catalog Number Availability Unit Your Price Price Per Qty