Would you like to log in again?
ALT Liquid Reagent Set, Pointe Scientific
Supplier: Pointe Scientific
HA926-610 A7526-150 A7526-625 A7526-450 OA926-487
10023-460EA 517.66 USD InStock
10023-460 10023-534 10022-978 10023-532 10023-536
ALT Liquid Reagent Set, Pointe Scientific
Assays Enzyme Assays
ALT Liquid Reagent Set is used for the quantitative determination of Alanine Aminotransferase in serum.
  • Catalyzes the transfer of the amino group from L-alanine to α-ketoglutarate
  • Simultaneous oxidation of NADH to NAD
  • Rate of decrease in absorbance is directly proportional to ALT activity
ALT Liquid Reagent Set is used for the quantitative determination of Alanine Aminotransferase in serum
The Olympus kit is for use on the Beckman Coulter AU 400, 480, and 640 analyzers and is for in vitro diagnostic use only. There are six 67 mL R1 Reagent bottles and five 17 mL R2 Reagent bottles in the Olympus kit.
Certifications: CE Compliant
Ordering information: Materials Required but not Provided:
The validity of the reaction should be monitored using control sera with known normal and abnormal ALT (SGPT) values (C7590/91/92).
Order Now
ORDER
Q and A
Product Questions and Answers
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.
Original Product:
Description Catalog Number Availability Unit Your Price Price Per Qty