Would you like to log in again?
Hemoglobin A1c Reagent Set, Pointe Scientific
Supplier: Pointe Scientific
OH546-40
10023-488EA 1190.77 USD InStock
10023-488
Hemoglobin A1c Reagent Set, Pointe Scientific
Clinical Diagnostic Supplies Clinical Reagents and Kits
For the quantitative determination of Hemoglobin A1c (HbA1c) in human blood
This method utilizes the interaction of antigen and antibody to directly determine the HbA1c in whole blood. Total hemoglobin and HbA1c have the same unspecific absorption rate to latex particles. When mouse antihuman HbA1c monoclonal antibody is added (R2), latex-HbA1c-mouse anti human HbA1c antibody complex is formed. Agglutination is formed when goat anti-mouse IgG polyclonal antibody interacts with the monoclonal antibody. The amount of agglutination is proportional to the amount of HbA1c absorbed on to the surface of latex particles. The amount of agglutination is measured as absorbance. The HbA1c value is obtained from a calibration curve.
The Olympus Hemoglobin A1c reagent is for use on the Beckman Coulter AU 400, 480, and 640 analyzers.
The determination of HbA1c is most commonly performed for the evaluation of glycemic control in diabetes mellitus. HbA1c values provide an indication of glucose levels over the preceding 4-8 weeks. A higher HbA1c value indicates poorer glycemic control. For in vitro diagnostic use only.
Certifications: CE Compliant, Health Canada (For OH546-40 Only)
Ordering information: Hemoglobin A1c calibrator set, control set, and Hemolysis Reagent are required but not provided (Provided in the Reagent Kit only).
Order Now
ORDER
ACCESSORIES
DOCUMENTATION
Q and A
Product Questions and Answers
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.
Original Product:
Description Catalog Number Availability Unit Your Price Price Per Qty