Would you like to log in again?
Lactate Dehydrogenase (Hitachi & Beckman) Reagent Set, Pointe Scientific
Supplier: Pointe Scientific
OL956-295 HL956-295
10023-492EA 607.25 USD InStock
10023-492 10022-958
Lactate Dehydrogenase (Hitachi & Beckman) Reagent Set, Pointe Scientific
Assays Enzyme Assays
The Lactate Dehydrogenase (Hitachi & Beckman) Reagent Set is for the quantitative kinetic determination of lactate dehydrogenase activity in serum.
  • Quantitative kinetic determination of lactate dehydrogenase activity in serum
  • Catalyzes the oxidation of lactate to pyruvate with simultaneous reduction of NAD to NADH
  • For in vitro diagnostic use only
The Lactate Dehydrogenase (Hitachi & Beckman) Reagent Set is for the quantitative kinetic determination of lactate dehydrogenase activity in serum
The Hitachi 295 mL reagent is for use on the Hitachi 917 analyzer only and for in vitro diagnostic use only. There are five 42 mL R1 Reagent bottles and five 17 mL R2 Reagent bottles in this kit.
The Hitachi 568 mL reagent is for use on Hitachi 717, 911, 912, and 914 analyzers and for in vitro diagnostic use only. There are four 100 mL R1 Reagent bottles and eight 21 mL R2 Reagent bottles in this kit.
The Olympus reagent is for use on the Beckman Coulter AU 400, 480, and 640 analyzers and is for in vitro diagnostic use only. There are five 42 mL R1 Reagent bottles and five 17 mL R2 Reagent bottles in the Olympus kit.
Certifications: CE Certified
Ordering information: Materials Required but not Provided:
Control sera with known normal and abnormal Lactate Dehydrogenase values.
Order Now
ORDER
Q and A
Product Questions and Answers
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.
Original Product:
Description Catalog Number Availability Unit Your Price Price Per Qty