Would you like to log in again?
HCG Reagent Set, Pointe Scientific
Supplier: Pointe Scientific
F1027-96
10023-510EA 398.1 USD InStock
10023-510
HCG Reagent Set, Pointe Scientific
Assays Enzyme Assays
The HCG reagent set is for the quantitative determination of Human Chorionic Gonadotropin (hCG) concentration in human serum.
  • Quantitative determination of Human Chorionic Gonadotropin (hCG) concentration in human serum
  • For in vitro diagnostic use only
  • Test is based on a solid phase enzyme-linked immunosorbent assay (ELISA)
The HCG reagent set is for the quantitative determination of Human Chorionic Gonadotropin (hCG) concentration in human serum. For in vitro diagnostic use only. The hCG quantitative test is based on a solid phase enzyme-linked immunosorbent assay (ELISA). The assay system utilizes a mouse monoclonal anti-α-hCG antibody for solid phase (microtiter wells) immobilization and another mouse monoclonal anti-β-hCG antibody in the antibody-enzyme (horseradish peroxidase) conjugate solution. The test specimen (serum) is added to the α-hCG antibody coated microtiter wells and incubated with the Zero Buffer at room temperature for 30 minutes. If hCG is present in the specimen, it will combine with the antibody on the wells. The wells are then washed to remove any residual test specimen, and β-hCG monoclonal antibody labeled with horseradish peroxidase (conjugate) is added. The conjugate will bind immunologically to the hCG on the wells, resulting in the hCG molecules being sandwiched between the solid phase and enzyme-linked antibodies. After incubation at room temperature for 15 minutes, the wells are washed with water to remove unbound-labeled antibodies. A solution of TMB Reagent is added and incubated at room temperature for 20 minutes, resulting in the development of a blue color. The color development is stopped with the addition of Stop Solution, and the color is changed to yellow and measured spectrophotometrically at 450nm. The concentration of hCG is directly proportional to the color intensity of the test sample.
Certifications: CE Certified
Ordering information: Materials Required but not Provided:
Quality control material (e.g., BioRad Lyphochek Control sera)
Order Now
ORDER
Q and A
Product Questions and Answers
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.
Original Product:
Description Catalog Number Availability Unit Your Price Price Per Qty