Would you like to log in again?
Alkaline Phosphatase Reagent Set, Pointe Scientific
Supplier: Pointe Scientific
A7516-625 HA916-492 A7516-450 A7516-150 OA916-302
10023-528EA 209.08 USD InStock
10023-528 10023-530 10022-976 10023-526 10023-458
Alkaline Phosphatase Reagent Set, Pointe Scientific
Assays Enzyme Assays
The Alkaline Phosphatase Reagent Set is used for the quantitative determination of alkaline phosphatase in human serum.
  • Quantitative determination of alkalin phosphatase in human serum
  • In vitro diagnostic use only
For the quantitative determination of alkaline phosphatase in human serum
The Hitachi 302 mL kit is for use on the Hitachi 917 analyzer only and for in vitro diagnostic use only. There are five 50 mL R1 Reagent bottles and four 13 mL R2 Reagent bottles in this kit.
The Hitachi 492 mL kit is for use on Hitachi 717, 911, 912, and 914 analyzers and for in vitro diagnostic use only. There are four 100 mL R1 Reagent bottles and four 23 mL R2 Reagent bottles in this kit.
Certifications: CE Compliant
Ordering information: Materials Required but not Provided:
The validity of the reaction should be monitored using control sera with known normal and abnormal ALP activities (C7590/91/92)
Order Now
ORDER
Q and A
Product Questions and Answers
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.
Original Product:
Description Catalog Number Availability Unit Your Price Price Per Qty