Would you like to log in again?
F-314 CHILLER 110V
Supplier: Buchi
Chiller^ F-314 Chiller 110V, Temperature range: -10 to +25 Deg C, Tank Volume: 6.5L, Flow Rate: 11 L\min, Pump Pressure: 1 bar, Power Consumptions (max.): 1500W, Voltage: 115 VAC +/- 10 %
ORDER
Expand All / Collapse All
Description VWR Catalog Number Supplier No Unit Price Quantity
F-314 CHILLER 110V 10023-990 11F31402 Each Retrieving
Generate Barcodes Barcode Label Format: 
Generate PDF Catalog Page
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.
Original Product:
Description Catalog Number Availability Unit Your Price Price Per Qty