Would you like to log in again?
Ugo Basile Dynamic Plantar, Stoelting
Supplier: Stoelting
57820
100231-404EA 12678.08 USD InStock
100231-404
Ugo Basile Dynamic Plantar, Stoelting
Animal Supplies Animal Environmental Enrichment
The ugo basile dynamic plantar aesthesiometer has been designed to automate the assessment of touch sensitivity on the plantar surface of rats or mice and comes ready to work with both mice and rats. Similar to electronic Von Frey or Plantar Von Frey instrument, but it differs from Von Frey devices as latency and actual force at the time of paw withdrawal reflex are automatically detected and recorded.
  • Movable touch-stimulator unit well proven in revious model
  • Large testing surface
  • Multifunction liquid-crystal graphic-display
  • New modular animal enclosure, offering from 3 to 12 spaces
The unit comes with a microprocessor controlled electronic unit, of new design provided with graphic display, internal memory for data storage, memory stick and optional printer.
DELTA 9-pin connector
85 - 264 V, 50 - 60 Hz
10.2 kg
Order Now
ORDER
Q and A
Product Questions and Answers
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.
Original Product:
Description Catalog Number Availability Unit Your Price Price Per Qty