Would you like to log in again?
R-300H 1L 24/40 C PG 110
Supplier: Buchi
Rotavapour^ Rotavapour, R-300H 1L 24/40 C PG 110, Lift System: Manual, Heating Bath: B-301 (20 - 95 Deg C, 1L), Standard Joint: SJ 24/40, Glass Assembly: Cold trap (C), Protective Coating: P+G safety coating, Voltage: 100 - 120V
ORDER
Expand All / Collapse All
Description VWR Catalog Number Supplier No Unit Price Quantity
R-300H 1L 24/40 C PG 110 10024-006 11R300112C102 Each Retrieving
Generate Barcodes Barcode Label Format: 
Generate PDF Catalog Page
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.
Original Product:
Description Catalog Number Availability Unit Your Price Price Per Qty