Would you like to log in again?
Scheduled maintenance of the site is planned from June 8th at 7:00 PM to June 9th at 5:00 PM ET. During this time, contract pricing and registration functionality will be unavailable. We appreciate your patience.
Compliance Gold® Occult Blood (iFOB) Tests, Germaine® Laboratories
Supplier: Germaine Laboratories
Compliance Gold®
74015 74030 74008 74014 74033 74034
10024-738BX 647.88 USD InStock
10024-738 10024-806 10024-808 10024-740 10024-742 10024-810
Compliance Gold® Occult Blood (iFOB) Tests, Germaine® Laboratories
Clinical Diagnostic Supplies Clinical Reagents and Kits
Compliance Gold® iFOB (immunological fecal occult blood) is a rapid chromatographic assay for the qualitative detection of occult blood in human stool samples
Compliance Gold® tests are packaged in various sizes to fit every need. Some versions allow for the patient to take the test home and return specimens via mail. Complete return mailers include a collection bottle, collection paper, specimen bag, and easy to follow instructions.
The full compliance verison includes a Compliance Monotoring System, during which a team member will provide a HIPAA compliant program that reminds patients to return collection kits.
These tests are used to determine gostrointestinal disorders such as colitis, colorectal cancer, diverticulitis, and polyps.
Certifications: CLIA waived.
Order Now
ORDER
DOCUMENTATION
Q and A
Product Questions and Answers
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.
Original Product:
Description Catalog Number Availability Unit Your Price Price Per Qty