Would you like to log in again?
Limited Volume Vial Insert, Kimble Chase
Supplier: DWK Life Sciences
60855-531 60855-631 60850-528 60850-629
10024-790CS 189.93 USD InStock
10024-790 10024-792 10024-794 10024-788
Limited Volume Vial Insert, Kimble Chase
Vials
Inserts are ideal for limited and micro volume applications.
  • Manufactured from clear borosilicate glass
Options are available for both standard and wide opening 12×32 autosampler type vials.
60850 series has a conical point design with polymer springs attached and residual volume of 1 µl. 60855 series has a flat bottom design and residual volume of 25 µl.
Order Now
ORDER
DOCUMENTATION
Q and A
Product Questions and Answers
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.
Original Product:
Description Catalog Number Availability Unit Your Price Price Per Qty