Would you like to log in again?
Febrile Antigens and Antisera, Germaine® Laboratories
Supplier: Germaine Laboratories
1002 1200 1303 1702 1402 1000 1503 1400 1600 1601 1501 1403 1100 1502 1201 1500 1611 1401 1606 1703 1202
10024-924EA 60.24 USD InStock
10024-924 10024-902 10024-912 10024-926 10024-904 10024-914 10024-906 10024-928 10024-916 10024-908 10024-918 10024-890 10024-892 10024-894 10024-896 10024-920 10024-898 10024-900 10024-922 10024-888 10024-910
Febrile Antigens and Antisera, Germaine® Laboratories
Clinical Diagnostic Supplies Clinical Reagents and Kits
Febrile Antigens are intended for the detection of bacterial agglutinins in serum associated with a bacterial infection
The slide test is recommended as a screening procedure to establish the presence or absence of homologous antibodies. The tube test is intended to establish antibody titer. The Febrile Controls are intended for use as positive and negative controls for these tests.
Order Now
ORDER
DOCUMENTATION
Q and A
Product Questions and Answers
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.
Original Product:
Description Catalog Number Availability Unit Your Price Price Per Qty