Would you like to log in again?
Gelatin Subbed Slides, Southern Biotech
Supplier: Southern Biotechnology
SLD01-CS SLD01-BX
100241-864CS 784.62 USD InStock
100241-864 100241-866
Gelatin Subbed Slides, Southern Biotech
Slides Microscopy Control Slides
Gelatin Subbed Slides are subbed with a solution of 0.1% gelatin and chromium potassium sulfate.
  • For improved section adhesion
  • Subbed with a solution of 0.1% gelatin and chromium potassium sulfate
  • Carefully cleaned 25 x 75 mm slides
SouthernBiotech’s Gelatin Subbed Slides have been carefully cleaned and subbed with a solution of 0.1% gelatin and chromium potassium sulfate. The coated slides measure 25 x 75mm, with a frosted end for marking. For optimal adhesion, sections less than 100μm in thickness are recommended.
Order Now
ORDER
Q and A
Product Questions and Answers
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.
Original Product:
Description Catalog Number Availability Unit Your Price Price Per Qty