Would you like to log in again?
ApoScreen® Annexin V Apoptosis Kit-FITC, Southern Biotechnology
Supplier: Southern Biotechnology
10010-02
100242-222EA 374 USD InStock
100242-222
ApoScreen® Annexin V Apoptosis Kit-FITC, Southern Biotechnology
Assays Cellular Assays
FITC ApoScreen® Annexin V Apoptosis Kit for use in flow cytometry and immunocytochemistry assays.
  • Contains Annexin V-FITC, Propidium Iodide (PI), and Annexin V Binding Buffer 10X
  • For detection of apoptosis in flow cytometry and immunocytochemistry assays
  • Differentiates apoptotic from necrotic cells
ApoScreen® Annexin V Apoptosis Kit-FITC (SB Cat. No. 10010-02) employs a fluorescein-conjugated Annexin V (Annexin V-FITC) in concert with propidium iodide (PI) to evaluate subpopulations of cells undergoing apoptosis. During the early stages of apoptosis, cells begin to display phosphatidylserine (PS) on the outer cell membrane where it is readily detectable by staining the cells with Annexin V-FITC. As the plasma membrane becomes increasingly permeable during the later stages of apoptosis, PI can readily move across the cell membrane and bind to cellular DNA, providing a means for identifying those cells that have lost membrane integrity through mechanisms including necrosis. When cells are double stained with Annexin V-FITC and PI, three different cell populations may be observed - (i) live cells that do not stain with either Annexin V-FITC or PI; (ii) necrotic cells that stain with both reagents; and (iii) apoptotic cells that stain with Annexin V-FITC only. Analysis may be performed on any flow cytometer equipped with a single laser using excitation at 488 nm. Alternatively, the cells can be examined with a fluorescence microscope equipped with filters for discrimination of FITC and rhodamine/PI.
Order Now
ORDER
Q and A
Product Questions and Answers
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.
Original Product:
Description Catalog Number Availability Unit Your Price Price Per Qty