Would you like to log in again?
ApoScreen® Propidium Iodide (PI), Southern Biotechnology
Supplier: Southern Biotechnology
10044-01
100242-250EA 85 USD InStock
100242-250
ApoScreen® Propidium Iodide (PI), Southern Biotechnology
Assays Cellular Assays
ApoScreen® Propidium Iodide (PI) for use in flow cytometry assays.
  • Ready-to-use cell viability dye
  • For detection of necrotic cells in flow cytometry and immunocytochemistry assays
  • Excitation/emission – 536/617 nm
Propidium Iodide (PI) is a ready-to-use cell viability dye for the exclusion of nonviable cells in flow cytometry analysis. The solution can be used in conjunction with fluorescein (FITC), cyanine-5 (CY5), and allophycocyanin (APC) conjugated Annexin V to distinguish cells that are in the early versus late stages of apoptosis.
Excitation/emission – 536/617 nm
Order Now
ORDER
Q and A
Product Questions and Answers
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.
Original Product:
Description Catalog Number Availability Unit Your Price Price Per Qty