Would you like to log in again?
ApoScreen® Annexin V Binding Buffer 10X, Southern Biotechnology
Supplier: Southern Biotechnology
10045-01
100242-252EA 57.54 USD InStock
100242-252
ApoScreen® Annexin V Binding Buffer 10X, Southern Biotechnology
Assays Cellular Assays
ApoScreen® Annexin V Binding Buffer 10X for use in flow cytometry assays.
  • For use with ApoScreen® Annexin V reagents
  • 10X formulation
  • Simply dH20 prior to use
ApoScreen® Annexin V Binding Buffer 10X has been formulated for flow cytometric labeling of apoptotic cells with ApoScreen® Annexin V reagents.
Order Now
ORDER
Q and A
Product Questions and Answers
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.
Original Product:
Description Catalog Number Availability Unit Your Price Price Per Qty