Would you like to log in again?
ISO 2431 Immersion Cup, Industrial Physics
Supplier: Industrial Physics
REVIEWS (0)
Total Ratings: 0
Avg. Ratings: out of 5
Sort all by:
VF2091 VF2222 VF2185 VF2093 VF2220 VF2090 VF2092 VF2224
10025-158EA 371.34 USD InStock
10025-158 10025-168 10025-166 10025-164 10025-162 10025-172 10025-160 10025-170
ISO 2431 Immersion Cup, Industrial Physics
Sample Containers General Purpose Containers
The ISO 2431 Immersion Cup is available in a range of titanium anodized aluminum or stainless steel formats
  • Each cup has a long handle, allowing it to be dipped by hand into a liquid container
  • Design of the cup and orifice eliminate hard to clean recesses
  • Cups are manufactured under continuing quality control procedures
  • Each cup features a unique, engraved serial number
These viscosity cups feature a fixed stainless steel nozzle (inner cavity) and handle. Inner dimensions are similar to ISO 2431. Ideal for measuring coatings and other fluids during application or production. Each immersion cup is supplied with a hard plastic storage case, with protective soft material on the interior.
Order Now
ORDER
ACCESSORIES
WRITE A REVIEW
REVIEWS (0)
Q and A
Product Questions and Answers
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.
Original Product:
Description Catalog Number Availability Unit Your Price Price Per Qty