Would you like to log in again?
VISCOSITY ACCESSORIES TE0027
Supplier: GREAT LAKES GAGES, LLC
Thermometer^ Precision Thermometer, Temperature Range: -49.9 to +149.9C, Resolution: 0.1C (F alterable), Accuracy: +/-0.5C between -49.9 to 99.9C, +/-C above 100C, Battery: MN2400 (AAA), Display: 15mm LCD, Dimensions: 19 x 52 x 155mm, Battery Lifetime: 8000 hours, Weight: 76 gram
ORDER
Expand All / Collapse All
Description VWR Catalog Number Supplier No Unit Price Quantity
VISCOSITY ACCESSORIES TE0027 10025-180 TE0027 Each Retrieving
Generate Barcodes Barcode Label Format: 
Generate PDF Catalog Page
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.
Original Product:
Description Catalog Number Availability Unit Your Price Price Per Qty