Would you like to log in again?
VISCOSITY ACCESSORIES VF2053
Supplier: GREAT LAKES GAGES, LLC
Visocity Calculator^ Visocity Calculator, indicative conversion table relating viscosity (in Cst) to flow time of different cups. Printed on the front is the No.4 complying with BS-NF-ASTM-Din and Zahn 2, on the back ISO nos 3-4-5-6 and Zahn 3 as well as the Gardner viscosimeter
ORDER
Expand All / Collapse All
Description VWR Catalog Number Supplier No Unit Price Quantity
VISCOSITY ACCESSORIES VF2053 10025-186 VF2053 Each Retrieving
Generate Barcodes Barcode Label Format: 
Generate PDF Catalog Page
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.
Original Product:
Description Catalog Number Availability Unit Your Price Price Per Qty