Would you like to log in again?
BAKER FILM APPLICATOR VF1527
Supplier: GREAT LAKES GAGES, LLC
Applicator^ Cylindrical film applicator with 4 application sides for applicating paint-films of 4 different pre-defined thicknesses, width 150mm, gaps 30/60/90120um, suitable for applying a host of different products onto flat and relatively solid substrates, not affected by acid/base elements
ORDER
Expand All / Collapse All
Description VWR Catalog Number Supplier No Unit Price Quantity
BAKER FILM APPLICATOR VF1527 10025-210 VF1527 Each Retrieving
Generate Barcodes Barcode Label Format: 
Generate PDF Catalog Page
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.
Original Product:
Description Catalog Number Availability Unit Your Price Price Per Qty