Would you like to log in again?
BAR FILM APPLICATOR SINGLE GAP VF1543
Supplier: GREAT LAKES GAGES, LLC
Applicator^ Bird-type film applicator with one flat edge, 4 for applying 1 pre-defined thicknesses, suitable for applying a host of different products onto flat and relatively solid substrates, high-grade SS unffected by acid/base element, Size: 50mm, 100um
ORDER
Expand All / Collapse All
Description VWR Catalog Number Supplier No Unit Price Quantity
BAR FILM APPLICATOR SINGLE GAP VF1543 10025-226 VF1543 Each Retrieving
Generate Barcodes Barcode Label Format: 
Generate PDF Catalog Page
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.
Original Product:
Description Catalog Number Availability Unit Your Price Price Per Qty