Would you like to log in again?
MICROSCOPE ILLUM HANDHELD 135X50X25 MM
Supplier: GREAT LAKES GAGES, LLC
Microscope^ Surface microscope with a built-in lamp for surface inspections, Magnification 40x, and equipped with a focus wheel to adjust the focus, a measuring reticle and an adjustable direct light source, Delivery in soft pouch, Dimensions: 135 x 50 x 25 mm, Weight: 110 grams
ORDER
Expand All / Collapse All
Description VWR Catalog Number Supplier No Unit Price Quantity
MICROSCOPE ILLUM HANDHELD 135X50X25 MM 10025-268 LD6205 Each Retrieving
Generate Barcodes Barcode Label Format: 
Generate PDF Catalog Page
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.
Original Product:
Description Catalog Number Availability Unit Your Price Price Per Qty