Would you like to log in again?
PH TEST 10 WATERPROOF
Supplier: GREAT LAKES GAGES, LLC
PH meter^ PH meter with large display, waterproof, equipped with automatic calibration with buffer recognition, accuracy of the meter is 0, 1 Ph. With automatic temperature compensation
ORDER
Expand All / Collapse All
Description VWR Catalog Number Supplier No Unit Price Quantity
PH TEST 10 WATERPROOF 10025-272 HI0019 Each Retrieving
Generate Barcodes Barcode Label Format: 
Generate PDF Catalog Page
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.
Original Product:
Description Catalog Number Availability Unit Your Price Price Per Qty