Would you like to log in again?
TQC DEWCHECK 4
Supplier: GREAT LAKES GAGES, LLC
Dewpoint Meter (DPM)/ Dewmeter^ Dewpoint Meter (DPM)/ Dewmeter, extremely versatile to measure and record all climate parameters required to treat surfaces, measures the relative humidity (%RH), ambient temperature (Ta) and surface temperature (Ts) and calculates the dewpoint temperature(Td)
ORDER
Expand All / Collapse All
Description VWR Catalog Number Supplier No Unit Price Quantity
TQC DEWCHECK 4 10025-274 DC7100 Each Retrieving
Generate Barcodes Barcode Label Format: 
Generate PDF Catalog Page
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.
Original Product:
Description Catalog Number Availability Unit Your Price Price Per Qty