Would you like to log in again?
TQC KARSTEN TUBE REPLACEMENT VERTICAL
Supplier: GREAT LAKES GAGES, LLC
Penetration Test^ Karsten Tube Penetration Test vertical replacement, for measuring the degree or water penetration into building materials such as concrete, stone and plaster. Consists of a glass tube filled with water, bonded to the test material with plastiline
ORDER
Expand All / Collapse All
Description VWR Catalog Number Supplier No Unit Price Quantity
TQC KARSTEN TUBE REPLACEMENT VERTICAL 10025-278 LI7505 Each Retrieving
Generate Barcodes Barcode Label Format: 
Generate PDF Catalog Page
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.
Original Product:
Description Catalog Number Availability Unit Your Price Price Per Qty