Would you like to log in again?
Komet, Magnetic Stirring Bars, Thermo Scientific
Supplier: Thermo Scientific
50087930 50087902 50087909 50087924 50087924 50087909 50087930 50087902
89030-778PK 210.69 USD InStock
CA89030-776 89030-778 89030-776 CA89030-782 CA89030-780 89030-781 CA89030-778 10025-290
Komet, Magnetic Stirring Bars, Thermo Scientific
Stirrers Magnetic Stirring Bars
Generate higher torque than conventional stirring bars with Komet Stir Bars.
  • Magnetic power two to three times higher than that of conventional bars
  • Magnetic properties remain for many years
Komet magnetic stirring bars made from SmCo (Samarium Cobalt) have a PTFE coating which means they can be sterilised. Particularly recommended for high speed stirring, liquids with a high viscosity and stirring in vessels where there is a large distance between stirrer and stirring bar, such as measuring cyliners.
Order Now
ORDER
Q and A
Product Questions and Answers
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.
Original Product:
Description Catalog Number Availability Unit Your Price Price Per Qty