Would you like to log in again?
Datalogging RH/Temperature Pen, Sper Scientific
Supplier: Sper Scientific
800013C 800013
10025-404EA 394.9 USD InStock
10025-404 10025-406
Datalogging RH/Temperature Pen, Sper Scientific
Data Loggers
Displays current relative humidity and temperature readings, and automatically logs air temperature, relative humidity, dew point, and wet bulb.
  • Records up to 32000 data points
  • Download the data by plugging the pen directly into a USB port
Datalogging can be programed to begin immediately, at a predetermined time, activated by the unit's push button, or to continually roll over after a set number of data points. Green and red led lights indicate datalogging and alarms.
Certifications: CE, RoHS certified.
Ordering information: Units are supplied with software, ½ AA battery, tripod screw, and a wrist strap. The Certified pen includes a N.I.S.T. traceable certificate of compliance. All units include a manufacturer's five-year warranty.
Order Now
ORDER
SPECIFICATIONS
Q and A
Product Questions and Answers
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.
Original Product:
Description Catalog Number Availability Unit Your Price Price Per Qty