Would you like to log in again?
Cohesive Bandages, DUKAL™ Corporation
Supplier: Dukal
REVIEWS (0)
Total Ratings: 0
Avg. Ratings: out of 5
Sort all by:
8016ASLF 8035DB 8025DB 8045T 8035T 8036TLF 8036ASLF 8026TLF 8016TLF 8015T 8045AS 8046ASLF 8025T 8015DB 8015W 8046TLF 8026ASLF
10025-418BX 41.28 USD InStock
10025-418 10025-416 10025-426 10025-358 10025-424 10025-368 10025-422 10790-474 10025-366 10025-420 10025-354 10025-430 10025-364 10025-362 10025-372 10025-360 10025-370
Cohesive Bandages, DUKAL™ Corporation
First Aid Supplies Wound Dressings
Provides consistent support and maintains compression. Can easily be torn by hand and is self adherent, no clips or fasteners are needed.
  • Lightweight, porous, and conforms well with movement
  • Available with natural rubber latex, or latex-free
Easy to apply and remove, cohesive bandages offer latex and latex-free formats. They make an excellent alternative to tape. Common applications include support wrap for strains or sprains, compression bandage, overwrap for splints, or overwrap for primary dressings.
Order Now
ORDER
DOCUMENTATION
WRITE A REVIEW
REVIEWS (0)
Q and A
Product Questions and Answers
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.
Original Product:
Description Catalog Number Availability Unit Your Price Price Per Qty