Would you like to log in again?
Dual Probe Digital Thermometer with Alarm, Thermco
Supplier: Thermco
ACC8100MATB
10025-442EA 126.26 USD InStock
10025-442
Dual Probe Digital Thermometer with Alarm, Thermco
Thermometers Digital Thermometers
Dual temperature thermometer to monitor freezer and refrigerator simultaneously, with min/max time and date stamp alarm.
  • Audible and visual alarms
  • 5mL vaccine bottle probes filled with bio-safe medium
Order Now
ORDER
DOCUMENTATION
Q and A
Product Questions and Answers
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.
Original Product:
Description Catalog Number Availability Unit Your Price Price Per Qty