Keep your session open?
Ending In 
Your shopping session has expired.
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

VWR® Standard Line Microcentrifuge Tubes, PP

Supplier: VWR

REVIEWS (0)

Total Ratings: 0
Avg. Ratings: out of 5

Sort all by:

VWR®
MTNA916 MTNA917 MTNA915 MTNA901 MTNA1019 MTNA903 MTNA910 MTNA1026 MTNA909 MTNA902 MTNA1018 MTNA908 MTNA1010 MTNA1025
10025-718CS 578.68 USD InStock
10025-718 10026-086 10025-728 10025-738 10025-716 10025-726 10025-736 10025-714 10025-724 10025-734 10025-722 10025-732 10025-720 10025-730
VWR® Standard Line Microcentrifuge Tubes, PP
Tubes Microcentrifuge Tubes
These Conical-Bottom Microcentrifuge tubes are made of super-clear, high-quality USP Class 6 gamma resistant polypropylene.

  • Available in three volumes: 0.5, 1.5, and 2.0 ml
  • Engraved graduations ensure accuracy
  • Flat and frosted caps surface together with smooth and frosted body surface
  • Clear and legible marking area provided
  • Tubes can be spun up to 25000×g
  • Autoclavable at 121 °C and freezable to –80 °C
  • DNase-free, RNase-free, and non-pyrogenic

They feature a positive seal design that withstands repeated cap closures and will not pop open during boiling. Tubes fit all standard rotors.

Certifications: TSE/BSE-free Certification.

Ordering information: Bagged tubes are packaged in durable and re-sealable (zip closure) bags. Inner packaging is individually labeled for lot-to-lot traceability.

Order Now
 

ORDER

SPECIFICATIONS

DOCUMENTATION

WRITE A REVIEW

REVIEWS (0)

Q and A

Product Questions and Answers

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty