Would you like to log in again?
Scheduled maintenance of the site is planned from June 8th at 7:00 PM to June 9th at 5:00 PM ET. During this time, contract pricing and registration functionality will be unavailable. We appreciate your patience.
VWR® Standard Line Sterile Racks for 15 and 50 ml Conical-Bottom Tubes, PP
Supplier: VWR
CTNA233 CTNA235 CTNA226 CTNA228 CTNA222 CTNA234 CTNA230 CTNA227 CTNA218 CTNA224 CTNA219 CTNA229 CTNA225 CTNA220 CTNA221 CTNA231
10026-302CS 348.8 USD InStock
10026-302 10026-312 10026-304 10026-314 10026-320 10026-066 10026-300 10026-310 10026-068 10026-064 10026-070 10025-704 10026-306 10026-316 10026-308 10026-318
VWR® Standard Line Sterile Racks for 15 and 50 ml Conical-Bottom Tubes, PP
Racks Tube Racks
Polypropylene racks are available in seven colors, and are also offered in an assorted color pack with one of each color: blue, green, orange, red, and yellow.
  • Available in a variety of colors
  • Suitable for plug or flat cap tubes
  • Polypropylene construction
  • During the freezing process, it is better to use plastic rack
  • During high-speed centrifugation, it is better to use suitable rotor adapters
Racks accommodate plug or flat cap tubes.
Certifications: TSE/BSE-free Certification.
Ordering information: All racks are sterilized by irradiation SAL10-6, and supplied RNase-free, DNase-free, and non-pyrogenic.
Order Now
ORDER
DOCUMENTATION
Q and A
Product Questions and Answers
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.
Original Product:
Description Catalog Number Availability Unit Your Price Price Per Qty