Would you like to log in again?
Microcentrifuge Tubes, Polypropylene, Greiner Bio-One
Supplier: Greiner bio-one
REVIEWS (0)
Total Ratings: 0
Avg. Ratings: out of 5
Sort all by:
722201 722261 616201 616283 667201 623201 742270 616281 717201 716201 717261 616261
82051-122CS 240.57 USD InStock
82051-122 10803-072 89217-356 82051-120 82050-514 89204-782 82050-504 82050-934 82051-160 10026-350 82051-124 82050-530
Microcentrifuge Tubes, Polypropylene, Greiner Bio-One
Tubes Microcentrifuge Tubes
Microcentrifuge tubes are manufactured from highly transparent, inert polypropylene resin that offers superior chemical resistance and thermal stability.
  • Polypropylene construction
  • Highly transparent and inert
  • Superior chemical and temperature resistance
  • Autoclavable
  • Available nonsterile or sterile
Color-coded tubes are available for easy and quick identification.
Tubes are designed to suit most analyzer systems, including Eppendorf®, Viatron, Roche®, and Beckman® systems.
All 1.5 ml tubes feature molded graduations. Ideal solution for secure, micro-volume liquid storage. Options for conical, skirted bottom available. Tubes are nontoxic and nonpyrogenic.
Order Now
ORDER
DOCUMENTATION
WRITE A REVIEW
REVIEWS (0)
Q and A
Product Questions and Answers
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.
Original Product:
Description Catalog Number Availability Unit Your Price Price Per Qty