Keep your session open?
Ending In 
Your shopping session has expired.
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

DL-Proline ≥98%, Powder

Supplier: Avantor
Synonyms: (±)-ProlineH-DL-Pro-OH

0256425GM
100264-922EA 18 USD InStock
100264-922
DL-Proline ≥98%, Powder
DL-Proline
  • White to off-white powder
  • Store at 0-8 °C


Formula: C₅H₉NO₂
MW: 115.13 g/mol
Melting Pt: 208 °C (dec.)
MDL Number: MFCD00005250
CAS Number: 609-36-9
ADR: ,
https://us-legacy.vwr.com/stibo/web/std.lang.all/56/29/21285629.jpg
Order Now

 

ORDER

DOCUMENTATION

Q and A

Product Questions and Answers

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty