Keep your session open?
Ending In 
Your shopping session has expired.
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

Octyl-3,4,5-trihydroxybenzoate ≥98%

Supplier: Avantor
Synonyms: Gallic acid octyl esterOctyl gallaten-Octyl gallateOctyl 3,4,5-trihydroxybenzoate

0249625GM
100266-310EA 55 USD InStock
100266-310
Octyl-3,4,5-trihydroxybenzoate ≥98%
Octyl 3,4,5-trihydroxybenzoate


Formula: C₁₅H₂₂O₅
MW: 282.34 g/mol
Melting Pt: 96…102 °C
MDL Number: MFCD00002197
CAS Number: 1034-01-1
ADR: ,
https://us-legacy.vwr.com/stibo/web/std.lang.all/57/10/21285710.jpg https://us-legacy.vwr.com/stibo/web/std.lang.all/57/11/21285711.jpg
Order Now

 

ORDER

DOCUMENTATION

Q and A

Product Questions and Answers

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty