Would you like to log in again?
VWR® Signature™ 3D Rotator Waver
Supplier: VWR
VWR® Signature™
80863626 80863627
12620-916EA 2713 USD InStock
12620-916 10027-108
VWR® Signature™ 3D Rotator Waver
Shakers and Mixers Rotating/Rocking Shakers
3-D rotator waver is ideal for blood samples, DNA extractions, blotting techniques, and general mixing of various size tubes.
  • Vertical and horizontal orbital motion
  • Digital display for speed, tilt angle, and time
  • Microprocessor control
  • Timer with audible alarm
  • Now with Best-in-Class, Exclusive 5-Year Warranty
  • Available with a NIST Traceable, ISO/IEC 17025 Accredited Calibration Certificate from Troemner
Microprocessor touch pad provides electronic angle adjustment which allows users to adjust rocking angle while unit is operating
Precise speed control provides smooth, low foaming, three dimensional "wave" motion from 1 to 30rpm. Designed for use in incubators, CO2 incubators, cold rooms (-10 to 60°C), 80% relative humidity, non-condensing environments. Microprocessor controls with LED displays provide accurate and repeatable results. Spill-resistant design channels fluids away from internal components. Base is constructed of durable aluminum for extra stability.
Audible and visual signals will activate when system detects an overloaded, unbalanced condition or obstruction of the tray. LED display will show elapsed time or, when programmed to user-defined limit, will count down to zero and shut off unit. Audible alarm will sound when time reaches zero. Optional alarm mute can be set from the touch pad control. Powerful, maintenance-free stepper motor handles heavy loads up to 2.3kg (5lbs.) and provides a long service life. Optional stacking tray attaches to platform for higher capacity applications.
For improved repeatability, onboard memory recalls last settings even when unit is turned off.
Certifications: RoHS certified. CE Marked, TUV listed. Calibrated Shakers include a Troemner ISO/IEC Accredited Calibration Certificate traceable to NIST. The ISO/IEC 17025 Accredited Calibration Certificate includes multiple data points within the speed range and time function with the associated uncertainties. Troemner is accredited by NVLAP under Laboratory Code 105013-0 to perform ISO/IEC 17025 calibrations.
Ordering information: Unit includes a nonskid mat for platform and a 234 cm (92") detachable, three-wire cord and plug. The 230V unit is supplied with a European-style plug.
Order Now
ORDER
SPECIFICATIONS
ACCESSORIES
DOCUMENTATION
Q and A
Product Questions and Answers
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.
Original Product:
Description Catalog Number Availability Unit Your Price Price Per Qty