Would you like to log in again?
VWR® Signature™ Incubating 3-D Rotator Waver
Supplier: VWR
VWR® Signature™
80863633 80863632
10027-114EA 5078 USD InStock
10027-114 12620-920
VWR® Signature™ Incubating 3-D Rotator Waver
Shakers and Mixers Shaking Incubators
Incubating 3-D Rotator Waver combines the unique vertical and horizontal wave motion with general-purpose incubation into one compact unit.
  • Vertical and Horizontal Orbital Motion
  • Digital Display for Speed, Temperature, Tilt Angle, and Time
  • Microprocessor Control
  • Timer with Audible Alarm
  • Compact, Benchtop Design
  • Now with Best-in-Class, Exclusive 5-Year Warranty
  • Available with a NIST Traceable, ISO/IEC 17025 Accredited Calibration Certificate from Troemner
Microprocessor PID temperature controller maintains precise control and can be set in 1° increments
Easy to use touch pad controls allow the user to set temperature, mixing speed, and tilt angle directly from the front panel. Ideal for cell cultures, hybridization procedures, hematology, and blotting techniques. Features microprocessor controls with LED display for repeatable results. Durable base is constructed of cast aluminum for extra stability.
Electronic angle adjustment allows user to easily adjust rocking angle while unit is operating. Precise speed control provides smooth, low-speed rocking motion down to 1rpm. For improved repeatability, onboard memory recalls last speed and angle settings even when unit is turned off. LED display will show elapsed time or, when programmed to user-defined limit, will count down to zero and shut off unit. Audible alarm will sound when time reaches zero. Optional alarm mute function can be set from the touch pad control.
LEXAN® polycarbonate lid permits viewing of samples from all sides without disturbing internal temperatures.
Certifications: RoHS certified. CSA listed. Calibrated Shakers include a Troemner ISO/IEC Accredited Calibration Certificate traceable to NIST. The ISO/IEC 17025 Accredited Calibration Certificate includes multiple data points within the speed range and time function with the associated uncertainties. Troemner is accredited by NVLAP under Laboratory Code 105013-0 to perform ISO/IEC 17025 calibrations.
Ordering information: Unit includes a 234 cm (92") detachable, three-wire cord and plug.
Order Now
ORDER
SPECIFICATIONS
ACCESSORIES
DOCUMENTATION
Q and A
Product Questions and Answers
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.
Original Product:
Description Catalog Number Availability Unit Your Price Price Per Qty