Would you like to log in again?
VWR® Microplate Shaker, 230 V
Supplier: VWR
VWR®
80863444 980131-C
10027-122EA 2942.54 USD InStock
10027-122 12620-928
VWR® Microplate Shaker, 230 V
Shakers and Mixers Microplate Shakers
Ideal for immunoassays and general microplate shaking, microplate shakers feature a four-place platform that holds either two or four microplates or deep well blocks.
  • 230 V model
  • Optimized for shaking microplates
  • Digital display for speed and time
  • Microprocessor control
  • Timer with audible alarm
  • Holds two or four microplates
  • Now with best-in-class, exclusive five-year warranty
  • Available with a NIST traceable, ISO/IEC 17025 accredited calibration certificate from troemner
Plates are firmly locked into place with retaining springs. Microprocessor variable speed control provides consistent, uniform mixing action. Ramping feature slowly increases speed to set point for added safety. Independent LED displays feature speed and time. Timer will display elapsed time, or, when programmed to user-defined limit, will count down to zero and shut off unit. Audible alarm sounds when time reaches zero. The triple eccentric drive features permanently lubricated ball bearings. The maintenance-free, brushless DC motor provides a long service life. Ideal for use in incubators, CO₂ incubators, or cold rooms; shaker accommodates a −10 to +60 °C (−14 to +140 °F), 80% relative humidity, non-condensing environment. Durable base is constructed of cast aluminum for extra stability.
Certifications: RoHS certified. CSA listed and CE Marked. Calibrated Shakers include a Troemner ISO/IEC Accredited Calibration Certificate traceable to NIST. The ISO/IEC 17025 Accredited Calibration Certificate includes multiple data points within the speed range and time function with the associated uncertainties. Troemner is accredited by NVLAP under Laboratory Code 105013-0 to perform ISO/IEC 17025 calibrations.
Ordering information: Units include a 2.3m (7.5') detachable, three-wire cord and plug. Optional microtube rack attaches to platform to hold 1.5 mL or 2 mL microtubes and has a capacity of 70 tubes. 230V, 50/60Hz models are also available; contact your Avantor sales representative for more information.
Caution: 230 V model. Corresponding models with voltage suitable for use in US (12620-926 or 10027-120) are also available, please see separate product page.
Order Now
ORDER
SPECIFICATIONS
DOCUMENTATION
Q and A
Product Questions and Answers
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.
Original Product:
Description Catalog Number Availability Unit Your Price Price Per Qty