Keep your session open?
Ending In 
Your shopping session has expired.
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

VWR® Thermal Shake Touch Thermoshaker with 1.5 ml Block, 230 V

Supplier: VWR
VWR®
80863734 80863735
10027-176EA 5869 USD InStock
10027-176 89232-910
VWR® Thermal Shake Touch Thermoshaker with 1.5 ml Block, 230 V
Block Heaters Shaking Block Heater/Coolers
The VWR® Thermal Shake Touch is designed for applications that require consistent and precise results.

  • 220 V model, not for use in USA
  • Intuitive 4.3" LCD touchscreen
  • Rapid heating
  • High-speed shaking ability
  • Internal memory stores five separate 5-step programs
  • Operational data can be exported via USB
  • Interchangeable blocks ideal for processing micro-tubes, conical tubes, cryo tubes, and microplates
  • Now with Best-in-Class, Exclusive 5-Year Warranty
  • Available with a NIST Traceable, ISO/IEC 17025 Accredited Calibration Certificate from Troemner

With heating and shaking capabilities, the Thermal Shake Touch uses interchangeable blocks to accommodate a wide variety of tubes and microplates

The low-profile design minimizes the unit’s footprint on the bench. The LCD touch screen enables faster setting of temperature, speed, and time, which can all be viewed at once. Display features on-screen help topics with operational tips, and the touch screen is compatible with rubber gloves used in labs. USB port can transfer information to a flash drive for data logging, program storage, and software updates. Program control capabilities allow user programmable operation for automated use plus storage of five separate 5-step programs. Adjustable temperature ramp rate feature separately defines the heating rate in increments of 0.5 °C/min. For maximum temperature accuracy, the single point calibration procedure allows the user to calibrate at six distinctly defined temperatures. Though the unit can process an array of sample sizes in various VWR Blocks (sold separately), the Thermal Shake Touch conveniently and effectively operates with non-VWR Blocks as well. The unit is equipped with a pulse mode feature for quick vortex applications.

The VWR® Thermal Shake Touch has numerous innovative safety features that support consistent operation and ensure effective workplace safety. Constructed from a high quality heat- and chemical-resistant polymer, the unit’s housing remains cool to the touch throughout normal operating temperatures.A maximum temperature limiting function ensures the temperature will not exceed user defined limits, allowing user control of temperature sensitive samples. A hot-top warning light will illuminate when the temperature reaches 40 °C, and will remain lit until the temperature drops below 40 °C. In timed mode, an alarm will sound if the timer reaches zero, or if set point temperature is reached. Additionally, the heater will automatically shut off if the unit recognizes an internal issue. The unit’s enhanced electronics, temperature sensor, and audible and visible alarms provide users with both safe and accurate results.

The easy-to-use, 4.3" color LCD touch screen allows users to save and visibly track progress through the live status bar for five user-defined programs, each with five individual steps. The unit’s enhanced electronics and temperature sensor provide accurate and dependable temperature settings across the operating range.

Certifications: RoHS certified. TUV listed. Calibration: Calibrated Shakers include a Troemner ISO/IEC Accredited Calibration Certificate traceable to NIST. The ISO/IEC 17025 Accredited Calibration Certificate includes multiple data points within the temperature and speed range and time function with the associated uncertainties. Troemner is accredited by NVLAP under Laboratory Code 105013-0 to perform ISO/IEC 17025 calibrations.

Ordering information: Units include a 1.5 ml Block (89232-916) "rack, and cover" and a 234 cm (92") detachable, 3-wire cord and plug (230V units include a Euro-type plug).

Caution: 220 V model, not for use in USA. Corresponding model with voltage suitable for use in US is also available, please see separate product page.

Order Now
 

ORDER

SPECIFICATIONS

ACCESSORIES

DOCUMENTATION

Q and A

Product Questions and Answers

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty