Would you like to log in again?
Please be aware that our e-commerce platform will be undergoing scheduled maintenance from 6:00 AM to 2:00 PM ET on May 11, 2024. During this time our site will be not available. We appreciate your patience.
VWR® Advanced Microplate Vortex Mixers
Supplier: VWR
80862971 80862972
10027-198EA 923.93 USD InStock
10027-198 89399-884
VWR® Advanced Microplate Vortex Mixers
Shakers and Mixers Vortex Mixers
The VWR® Advanced Microplate Vortex Mixer is ideal for applications that demand repeatable results.
  • Designed for continuous duty
  • Designed for shaking microplates or tubes
  • LED displays for speed and time
  • Now with Best-in-Class, Exclusive 5-Year Warranty
  • Available with a NIST Traceable, ISO/IEC 17025 Accredited Calibration Certificate from Troemner
Mixer features touchpad controls and LED displays for accurate speed (rpm) and time (minutes and seconds) results
Touch/Standby/On rocker switch. LED displays for Speed/Time. Up/Down buttons for set-point controls.
Microprocessor control maintains set speed for strong, consistent mixing action. Timer will display elapsed time or when programmed to a user defined time limit, the unit will shut off when time reaches zero. Choose from two modes of operation: "Touch" mode which activates mixing when the cup head is depressed or "On" when using any of the accessory attachments for continuous operation. Continous duty rating.
Certifications: RoHS certified. TUV listed. CSA listed.230V units are CE marked. Calibrated Vortex Mixers include a Troemner ISO/IEC Accredited Calibration Certificate traceable to NIST. The ISO/IEC 17025 Accredited Calibration Certificate includes multiple data points within the speed range and time function with the associated uncertainties. Troemner is accredited by NVLAP under Laboratory Code 105013-0 to perform ISO/IEC 17025 calibrations.
Ordering information: Mixer includes a cup head, single microplate holder, and a 234 cm (92") detachable, 3-wire cord and plug.
Order Now
ORDER
SPECIFICATIONS
ACCESSORIES
DOCUMENTATION
Q and A
Product Questions and Answers
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.
Original Product:
Description Catalog Number Availability Unit Your Price Price Per Qty